Learn More
Accela Chembio Inc 3-fluoro-2-methylbenzaldehyde | 5g | 147624-13-3 | MFCD00075256 | 97+% | D: 1.16 | Shelf Life: 540 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50353607
Encompass_Preferred
3-fluoro-2-methylbenzaldehyde | 5g | 147624-13-3 | MFCD00075256 | 97+% | D: 1.16 | Shelf Life: 540 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-353-607 Supplier Accela Chembio Inc Supplier No. SY0030675G
3-fluoro-2-methylbenzaldehyde | 5g | 147624-13-3 | MFCD00075256 | 97+% | D: 1.16 | Shelf Life: 540 Days | Light Sensitive/nitrogen Or Argon/+4