Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-2-nitropyridine, 96%

Catalog No. AAH3381306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3381306 5 g
AAH3381303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3381306 Supplier Thermo Scientific Chemicals Supplier No. H3381306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 54231-35-5 | C5H3FN2O2 | 142.089 g/mol

A specialty product for proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A specialty product for proteomics research applications.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54231-35-5
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.089
MDL Number MFCD04114127
InChI Key IJVFHCSUEBAAOZ-UHFFFAOYSA-N
Synonym 2-nitro-3-fluoropyridine, 3-fluoro-2-nitro-pyridine, pyridine, 3-fluoro-2-nitro, pubchem2185, 3-fluor-2-nitropyridin, pyfn51, 3-fluoro 2-nitro pyridine, pyridine,3-fluoro-2-nitro, pyridine, 3-fluoro-2-nitro-9ci
PubChem CID 2762802
IUPAC Name 3-fluoro-2-nitropyridine
SMILES C1=CC(=C(N=C1)[N+](=O)[O-])F

Spécifications

Melting Point 30°C to 35°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 96%
Chemical Name or Material 3-Fluoro-2-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.