Learn More
Medchemexpress LLC 3-fluoro-2-(pyrimidin-2-yl)phenyl-7-azabicyclo[2.2.1]heptan-7-yl methanone | 1628843-99-1 | 98.1% | C22H18F4N6O | 10MG
SDP
Catalog No. 5000259802
Encompass_Preferred
JNJ-54717793 is a research-stage, brain-penetrant selective orexin-1 receptor (OX1R) antagonist developed by Janssen Research & Development and characterized for preclinical pharmacology and behavioral models. It is described in peer-reviewed studies and is supplied to researchers for laboratory use.
  • selective orexin-1 receptor antagonist with high affinity.
  • brain-penetrant pharmacokinetic profile suitable for in vivo studies.
  • high reported purity (98.1%).
  • characterized in peer-reviewed pharmacology studies.
  • supplied for research use only, not for human or veterinary use.
Catalog No. 50-002-59802 Supplier Medchemexpress LLC Supplier No. HY13418810MG
JNJ-54717793 is a research-stage, brain-penetrant selective orexin-1 receptor (OX1R) antagonist developed by Janssen Research & Development and characterized for preclinical pharmacology and behavioral models. It is described in peer-reviewed studies and is supplied to researchers for laboratory use.
  • selective orexin-1 receptor antagonist with high affinity.
  • brain-penetrant pharmacokinetic profile suitable for in vivo studies.
  • high reported purity (98.1%).
  • characterized in peer-reviewed pharmacology studies.
  • supplied for research use only, not for human or veterinary use.