Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluoro-2-(tributylstannyl)pyridine | 573675-60-2 | 386.14 | C17H30FNSn | 5 G
SDP

Catalog No. 5000210430
Encompass_Preferred

3-Fluoro-2-(tributylstannyl)pyridine is an organostannane research reagent used as a building block in organic synthesis and biochemical assays. It is supplied as a liquid and intended for research use only; recommended storage conditions are provided for both the pure material and solutions.

  • Organostannane reagent for organic synthesis and biochemical assays.
  • Molecular formula C17H30FNSn; molecular weight 386.14.
  • CAS number 573675-60-2.
  • Appearance colorless to light yellow liquid.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or clinical use.
  • Solubility not specified.

Catalog No. 50-002-10430 Supplier Medchemexpress LLC Supplier No. HYW0708135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Fluoro-2-(tributylstannyl)pyridine is an organostannane research reagent used as a building block in organic synthesis and biochemical assays. It is supplied as a liquid and intended for research use only; recommended storage conditions are provided for both the pure material and solutions.

  • Organostannane reagent for organic synthesis and biochemical assays.
  • Molecular formula C17H30FNSn; molecular weight 386.14.
  • CAS number 573675-60-2.
  • Appearance colorless to light yellow liquid.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or clinical use.
  • Solubility not specified.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.