Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluoro-2-(tributylstannyl)pyridine | 573675-60-2 | 386.14 g/mol | C17H30FNSn | 500 MG
SDP

Catalog No. 5000210250
Encompass_Preferred

3-fluoro-2-(tributylstannyl)pyridine is an organotin-substituted pyridine provided as a research reagent and synthetic intermediate. It is identified by CAS number 573675-60-2 and has molecular formula C17H30FNSn (molecular weight 386.14 g/mol).

  • Organotin-substituted pyridine used as a synthetic building block
  • Molecular formula C17H30FNSn
  • Molecular weight 386.14 g/mol
  • CAS number 573675-60-2
  • Available in milligram-scale pack sizes (250 mg, 500 mg, 1 g, 5 g)
  • For research use only

Catalog No. 50-002-10250 Supplier Medchemexpress LLC Supplier No. HYW070813500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-fluoro-2-(tributylstannyl)pyridine is an organotin-substituted pyridine provided as a research reagent and synthetic intermediate. It is identified by CAS number 573675-60-2 and has molecular formula C17H30FNSn (molecular weight 386.14 g/mol).

  • Organotin-substituted pyridine used as a synthetic building block
  • Molecular formula C17H30FNSn
  • Molecular weight 386.14 g/mol
  • CAS number 573675-60-2
  • Available in milligram-scale pack sizes (250 mg, 500 mg, 1 g, 5 g)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.