Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluoro-2-(tributylstannyl)pyridine | 573675-60-2 | MFCD09265496 | 95.0% | 386.14 | C17H30FNSn | 250 MG
SDP

Catalog No. 5000209481
Encompass_Preferred

3-Fluoro-2-(tributylstannyl)pyridine is an organotin-substituted pyridine reagent used as an intermediate in organic synthesis and cross-coupling research applications. It is supplied as a colorless to light yellow liquid and is offered in laboratory pack sizes for research use.

  • Organotin-substituted pyridine reagent
  • Molecular weight 386.14
  • Chemical formula C17H30FNSn
  • Cas number 573675-60-2
  • Colorless to light yellow liquid
  • Available in small laboratory pack sizes (e.g., 250 MG)

Catalog No. 50-002-09481 Supplier Medchemexpress LLC Supplier No. 250MGHYW070813
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Fluoro-2-(tributylstannyl)pyridine is an organotin-substituted pyridine reagent used as an intermediate in organic synthesis and cross-coupling research applications. It is supplied as a colorless to light yellow liquid and is offered in laboratory pack sizes for research use.

  • Organotin-substituted pyridine reagent
  • Molecular weight 386.14
  • Chemical formula C17H30FNSn
  • Cas number 573675-60-2
  • Colorless to light yellow liquid
  • Available in small laboratory pack sizes (e.g., 250 MG)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.