Learn More
Selleck Chemical LLC 3 -Fluoro-3 -deoxythymidine
SDP
Catalog No. 5000001832
Encompass_Preferred
3 -Fluoro-3 -deoxythymidine (Alovudine CL 184824 FddThd FLT MIV-310) is a potent inhibitor of polymerase and reverse transcriptase that can be used in the treatment of HIV infection 3 -Fluoro-3 -deoxythymidine (Alovudine) is also a marker of DNA synthesis that can be used as an early response biomarker in the chemotherapy of pancreatic cancer
Catalog No. 50-000-01832 Supplier Selleck Chemical LLC Supplier No. S684825MG
3 -Fluoro-3 -deoxythymidine (Alovudine CL 184824 FddThd FLT MIV-310) is a potent inhibitor of polymerase and reverse transcriptase that can be used in the treatment of HIV infection 3 -Fluoro-3 -deoxythymidine (Alovudine) is also a marker of DNA synthesis that can be used as an early response biomarker in the chemotherapy of pancreatic cancer