Learn More
Medchemexpress LLC 3-Fluoro-4-formylphenylboronic acid | 248270-25-9 | 167.93 | 10 G
SDP
Catalog No. 5000447839
Encompass_Preferred
3-Fluoro-4-formylphenylboronic acid is a biochemical reagent suitable for life science related research. It can be used as a biological material or organic compound. For research use only.
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-004-47839 Supplier Medchemexpress LLC Supplier No. HYW00802710G
3-Fluoro-4-formylphenylboronic acid is a biochemical reagent suitable for life science related research. It can be used as a biological material or organic compound. For research use only.
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month