Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-iodobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. F10931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10931G Supplier TCI America Supplier No. F10931G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 887266-99-1
Molecular Formula C7H3FIN
Molecular Weight (g/mol) 247.011
MDL Number MFCD07782076
InChI Key NPKQMCCUPXZXFI-UHFFFAOYSA-N
Synonym benzonitrile, 3-fluoro-4-iodo, benzonitrile,3-fluoro-4-iodo, 4-cyano-2-fluoroiodobenzene, acmc-20aiba, pubchem4781, 5-fluoro-4-iodobenzonitrile, 3-fluoro-4-iodo benzonitrile, 3-fluoro-4-iodo-benzonitrile, 3-fluoro-4-iodobenzonitrile3, ksc661e6h
PubChem CID 24721521
IUPAC Name 3-fluoro-4-iodobenzonitrile
SMILES C1=CC(=C(C=C1C#N)F)I

Specifications

Melting Point 116°C
Quantity 1 g
Formula Weight 247.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-iodobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.