Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methoxybenzyl bromide, 98%

Catalog No. p-7028711
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26116MD 250 mg
AAH2611603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26116MD Supplier Thermo Scientific Chemicals Supplier No. H26116MD
Only null left
Add to Cart

CAS: 331-61-3 | C8H8BrFO | 219.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-61-3
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00671768
InChI Key KLWYYWVSFYRPLM-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzyl bromide, 4-bromomethyl-2-fluoro-1-methoxybenzene, 3-fluoro-4-methoxybenzylbromide, 4-bromomethyl-2-fluoro-1-methoxy-benzene, benzene, 4-bromomethyl-2-fluoro-1-methoxy, pubchem4923, acmc-1ckmo, 4-bromomethyl-2-fluoroanisole
PubChem CID 2774553
IUPAC Name 4-(bromomethyl)-2-fluoro-1-methoxybenzene
SMILES COC1=CC=C(CBr)C=C1F

Specifications

Melting Point 44°C to 46°C
Odor Pungent
Quantity 250 mg
UN Number UN3261
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.