Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F08765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0876-5G 5 g
F08761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0876-5G Supplier TCI America Supplier No. F08765G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 149507-26-6
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD00807404
InChI Key IILGLPAJXQMKGQ-UHFFFAOYSA-N
PubChem CID 2782194
IUPAC Name (3-fluoro-4-methoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OC)F)(O)O

Specifications

Melting Point 198°C
Quantity 5 g
Synonym 3-fluoro-4-methoxybenzeneboronic acid, 3-fluoro-4-methoxyphenyl boronic acid, 3-fluoro-4-methoxyphenyl boranediol, 3-fluoro-4-methoxyphenylboronicacid, 4-methoxy-3-fluorophenylboronic acid
Formula Weight 169.95
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.