Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-methylbenzenesulfonyl chloride, 97%

Catalog No. p-7047614
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1951603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1951603 Supplier Thermo Scientific Chemicals Supplier No. L1951603
Only null left
Add to Cart

CAS: 90260-13-2 | C7H6ClFO2S | 208.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90260-13-2
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD03094389
InChI Key YDUHIMCRFRIVFI-UHFFFAOYSA-N
Synonym 3-fluoro-4-methylbenzene-1-sulfonyl chloride, 3-fluoro-4-methylbenzenesulfonylchloride, 3-fluoro-4-methylbenzenesulphonyl chloride, 3-fluoro-4-methyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methyl, 3-fluoro-p-toluenesulphonyl chloride, 3-fluoro-p-toluenesulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methyl-9ci, chloro 3-fluoro-4-methylphenyl sulfone, pubchem11718
PubChem CID 2778600
IUPAC Name 3-fluoro-4-methylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)F

Specifications

Melting Point 46°C to 50°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.