Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 97%, Thermo Scientific Chemicals

Catalog No. p-4812942
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446510010 1 g Glass bottle
AC446510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446510010 Supplier Thermo Scientific Chemicals Supplier No. 446510010
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 211108-50-8 | C10H16FNO3 | 217.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211108-50-8
Molecular Formula C10H16FNO3
Molecular Weight (g/mol) 217.24
InChI Key JZNWQLLPLOQGOI-UHFFFAOYSA-N
Synonym 1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
PubChem CID 10560711
IUPAC Name tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C(C1)F

Specifications

Melting Point 75°C to 77°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 217.24
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.