Learn More
Medchemexpress LLC 3-fluoro-5-(2-pyridinyl)benzoic acid, 2-[(2-fluorophenyl)sulfonyl]hydrazide | 2055397-28-7 | MFCD31690337 | >=98% | 389.4 g/mol | C18H13F2N3O3S | 25 MG
SDP
Catalog No. 5000248335
Encompass_Preferred
WM-1119 is a small-molecule inhibitor of lysine acetyltransferase 6A (KAT6A/MOZ) used as a biochemical probe in epigenetics and oncology research. It exhibits potent biochemical activity (KAT6A IC50 = 6.3 nM) and demonstrated cellular activity (EMRK1184 B-cell lymphoma IC50 = 0.25 μM). Key identifiers: CAS 2055397-28-7; formula C18H13F2N3O3S; molecular weight 389.4 g/mol; purity ≥98%.
  • Potent KAT6A inhibition (IC50 = 6.3 nM).
  • Demonstrated cellular activity in lymphoma cells (IC50 = 0.25 μM).
  • High purity supports reproducible biochemical assays (≥98%).
  • Well-defined molecular properties for characterization.
  • Suitable as a research chemical probe in epigenetic studies.
Catalog No. 50-002-48335 Supplier Medchemexpress LLC Supplier No. HY10205825MG
WM-1119 is a small-molecule inhibitor of lysine acetyltransferase 6A (KAT6A/MOZ) used as a biochemical probe in epigenetics and oncology research. It exhibits potent biochemical activity (KAT6A IC50 = 6.3 nM) and demonstrated cellular activity (EMRK1184 B-cell lymphoma IC50 = 0.25 μM). Key identifiers: CAS 2055397-28-7; formula C18H13F2N3O3S; molecular weight 389.4 g/mol; purity ≥98%.
  • Potent KAT6A inhibition (IC50 = 6.3 nM).
  • Demonstrated cellular activity in lymphoma cells (IC50 = 0.25 μM).
  • High purity supports reproducible biochemical assays (≥98%).
  • Well-defined molecular properties for characterization.
  • Suitable as a research chemical probe in epigenetic studies.