Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-iodoaniline, 96%

Catalog No. AAH3445703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3445703 1 g
AAH3445706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3445703 Supplier Thermo Scientific Chemicals Supplier No. H3445703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 660-49-1 | C6H5FIN | 237.016 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-49-1
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD04116466
InChI Key HTEPMXLKKDKDAQ-UHFFFAOYSA-N
Synonym 5-fluoro-3-iodoaniline, 3-fluoro-5-iodo-phenylamine, pubchem4458, 3-fluoro-5-iodo aniline, 3-fluoro-5-iodophenylamine, 5-fluoro-3-iodophenylamine, benzenamine,3-fluoro-5-iodo, benzenamine, 3-fluoro-5-iodo
PubChem CID 2758986
IUPAC Name 3-fluoro-5-iodoaniline
SMILES C1=C(C=C(C=C1F)I)N

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 237.02
Percent Purity 96%
Chemical Name or Material 3-Fluoro-5-iodoaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.