Learn More
Sigma Aldrich (3-fluoro-5-isopropoxyphenyl)boronic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 4-hydroxy-2-butanone is an important pharmaceutical intermediate. gas phase reaction of the oh radicals with 4-hydroxy-2-butanone has been investigated by absolute rate method.4-hydroxy-2-butanone has been used in the synthesis of:
- 3-buten-2-one via dehydration over anatase-tio2 catalyst
- 4-methyl-5-hydroxymethylthiazole
- (+/-)-lineatin
- Mevalonic acid lactone
- Verrucarin
- α-hydroxylactones
Specifications
Specifications
| Molecular Formula | C9H12BFO3 |
| Linear Formula | C9H12BFO3 |
| MDL Number | MFCD07363781 |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 198 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.