Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-(trifluoromethyl)benzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. F11315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11315G 5 g
F11311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11315G Supplier TCI America Supplier No. F11315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 239087-09-3
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.026
MDL Number MFCD00061175
InChI Key LCVWAXYGYMZJER-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzyl bromide, 1-bromomethyl-3-fluoro-5-trifluoromethyl benzene, 3-fluoro-5-trifluoromethylbenzyl bromide, 3-fluoro-5-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-3-fluoro-5-trifluoromethyl, acmc-1cdjv, 3-trifluoromethyl-5-fluorobenzyl bromide, 3-fluoro-5-trifluoromethyl-benzyl bromide, 5-bromomethyl-3-fluoro-1-trifluoromethyl benzene
PubChem CID 2737575
IUPAC Name 1-(bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)F)CBr

Specifications

Color Yellow
Boiling Point 43°C
Quantity 5 g
UN Number 3265
Formula Weight 257.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.