Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluoro-DL-phenylalanine | 456-88-2 | MFCD00004273 | 98.0% | 183.18 | C9H10FNO2 | 10 G
SDP

Catalog No. 5000302295
Encompass_Preferred

3-fluoro-DL-phenylalanine is a fluorinated phenylalanine derivative supplied as a solid for research use only. It is characterized by a defined molecular weight and purity, and is suitable as a building block or intermediate in synthetic and medicinal chemistry research.

  • Phenylalanine derivative
  • CAS number 456-88-2
  • Molecular formula C9H10FNO2
  • Molecular weight 183.18
  • Purity 98.0%
  • Appearance solid, white to off-white
  • Available in multiple sizes including 10 g
  • For research use only

Catalog No. 50-003-02295 Supplier Medchemexpress LLC Supplier No. HYW01633910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-fluoro-DL-phenylalanine is a fluorinated phenylalanine derivative supplied as a solid for research use only. It is characterized by a defined molecular weight and purity, and is suitable as a building block or intermediate in synthetic and medicinal chemistry research.

  • Phenylalanine derivative
  • CAS number 456-88-2
  • Molecular formula C9H10FNO2
  • Molecular weight 183.18
  • Purity 98.0%
  • Appearance solid, white to off-white
  • Available in multiple sizes including 10 g
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.