Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-L-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. F0272100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0272100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0272100MG Supplier TCI America Supplier No. F0272100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19883-77-3
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00066450
InChI Key VWHRYODZTDMVSS-QMMMGPOBSA-N
Synonym (S)-2-Amino-3-(3-fluorophenyl)propionic Acid, H-Phe(3-F)-OH
PubChem CID 716315
ChEBI CHEBI:77957
IUPAC Name (2S)-2-amino-3-(3-fluorophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)F)CC(C(=O)O)N

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 183.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-L-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.