Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-p-cresol 98.0+%, TCI America™
SDP

Catalog No. F04811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04811G 1 g
F04815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04811G Supplier TCI America Supplier No. F04811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-78-8
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00143164
InChI Key GJOOCAXPERKNMN-UHFFFAOYSA-N
Synonym 3-fluoro-p-cresol, 3-fluoro-4-methyl-phenol, 2-fluoro-4-hydroxytoluene, phenol, 3-fluoro-4-methyl, pubchem1499, 3-fluoro-4-methyl phenol, acmc-209k1q, 3-fluoranyl-4-methyl-phenol, ksc494a2d
PubChem CID 2774613
IUPAC Name 3-fluoro-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)F

Specifications

Boiling Point 196°C
Quantity 1 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Fluoro-p-cresol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.