Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-p-tolunitrile 98.0+%, TCI America™
SDP

Catalog No. F04245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04245G Supplier TCI America Supplier No. F04245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 170572-49-3
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00153174
InChI Key KUQQONVKIURIQU-UHFFFAOYSA-N
Synonym 4-cyano-2-fluorotoluene, 3-fluoro-p-tolunitrile, 2-fluoro-4-cyanotoluene, benzonitrile, 3-fluoro-4-methyl, 4-methyl-3-fluorobenzonitrile, 3-fluoro-4-methyl-benzonitrile, 3-fluoro-4-methylbenzenecarbonitrile, benzonitrile, 3-fluoro-4-methyl-9ci, ncr cf d1, pubchem1557
PubChem CID 585140
IUPAC Name 3-fluoro-4-methylbenzonitrile
SMILES CC1=CC=C(C=C1F)C#N

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-p-tolunitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.