Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Fluoro-propylamine hydrochloride

Catalog No. CDS007794 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007794 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007794 Supplier Sigma Aldrich Supplier No. CDS007794250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-cyclohepten-1-one is an α,α-enone and its regioselective reaction with allyl indium reagent in the presence of tmscl has been reported
  • It reacts with bu2zn in the presence of catalytic amounts of cu(otf)2 and ch2-bridged azolium salts to give (s)-3-butylcycloheptanone

Specifications

CAS 64068-31-1
Molecular Formula C3H9ClFN
Linear Formula C3H9ClFN
Quantity 10 g
MDL Number MFCD07369956
Molecular Weight (g/mol) 113.56
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.