Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Fluoroacetanilide 96.0+%, TCI America™
SDP

Catalog No. F030225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F030225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F030225G Supplier TCI America Supplier No. F030225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 351-28-0
Molecular Formula C8H8FNO
Molecular Weight (g/mol) 153.156
MDL Number MFCD00017917
InChI Key AQLLDCFUQXGLHM-UHFFFAOYSA-N
Synonym 3'-fluoroacetanilide, n-3-fluorophenyl acetamide, 3-fluoroacetanilide, m-fluoroacetanilide, 3-acetamido-1-fluorobenzene, acetanilide, 3'-fluoro, 1-fluoro-3-acetamidobenzene, acetamide, n-3-fluorophenyl, acmc-209idd, n-acetyl-3-fluoroaniline
PubChem CID 9594
IUPAC Name N-(3-fluorophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)F

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 25 g
Formula Weight 153.16
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Fluoroacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.