Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Fluorobenzaldehyde

Catalog No. F500510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F500510G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F500510G Supplier Sigma Aldrich Supplier No. F500510G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Fluorobenzaldehyde

Specifications

CAS 456-48-4
Density 1.17 g/mL (at 25°C (literature))
Boiling Point 66°C to 68°C (20 mmHg)
Molecular Formula C7H5FO
Linear Formula FC6H4CHO
Refractive Index n20/D 1.518 (literature)
MDL Number MFCD00003348
Quantity 1 g
Molecular Weight (g/mol) 124.11
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.