Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluorobutan-1-amine hydrochloride | 1780799-10-1 | MFCD28538986 | 95.0% | 127.59 | C4H11ClFN | 250 MG
SDP

Catalog No. 5000208887
Encompass_Preferred

3-Fluorobutan-1-amine hydrochloride is a chemical intermediate used in the synthesis of active pharmaceutical compounds. It is supplied as a hydrochloride salt and is provided as an off-white to light yellow solid with a molecular weight of 127.59 g/mol and a typical purity of 95.0% (NMR). Store sealed at room temperature, protected from moisture and light; solutions are stable at -80°C for 2 years or -20°C for 1 year.

  • Hydrochloride salt form
  • Off-white to light yellow solid
  • Purity 95.0% (NMR)
  • Molecular formula C4H11ClFN
  • CAS number 1780799-10-1
  • Intended as a drug intermediate for synthesis
  • Storage: RT sealed, away from moisture and light; in solvent -80°C (2 years) or -20°C (1 year)
  • Available in small research quantities such as 250 MG

Catalog No. 50-002-08887 Supplier Medchemexpress LLC Supplier No. 250MGHYW049770
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Fluorobutan-1-amine hydrochloride is a chemical intermediate used in the synthesis of active pharmaceutical compounds. It is supplied as a hydrochloride salt and is provided as an off-white to light yellow solid with a molecular weight of 127.59 g/mol and a typical purity of 95.0% (NMR). Store sealed at room temperature, protected from moisture and light; solutions are stable at -80°C for 2 years or -20°C for 1 year.

  • Hydrochloride salt form
  • Off-white to light yellow solid
  • Purity 95.0% (NMR)
  • Molecular formula C4H11ClFN
  • CAS number 1780799-10-1
  • Intended as a drug intermediate for synthesis
  • Storage: RT sealed, away from moisture and light; in solvent -80°C (2 years) or -20°C (1 year)
  • Available in small research quantities such as 250 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.