Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluorobutan-1-amine hydrochloride | 1780799-10-1 | MFCD28538986 | 95.0% | 127.59 g/mol | C4H11ClFN | 100 MG
SDP

Catalog No. 5000210567
Encompass_Preferred

3-fluorobutan-1-amine hydrochloride is a fluorinated primary amine supplied as the hydrochloride salt and used as a chemical intermediate for the synthesis of active organic compounds.

  • Primary amine hydrochloride salt.
  • Fluorinated C4 building block for synthesis.
  • CAS number 1780799-10-1.
  • Molecular weight 127.59 g/mol.
  • Purity 95.0% (as reported on certificate of analysis).
  • Solid; store sealed at room temperature, away from moisture and light.
  • Commonly supplied in small laboratory pack sizes such as 100 mg.

Catalog No. 50-002-10567 Supplier Medchemexpress LLC Supplier No. HYW049770100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-fluorobutan-1-amine hydrochloride is a fluorinated primary amine supplied as the hydrochloride salt and used as a chemical intermediate for the synthesis of active organic compounds.

  • Primary amine hydrochloride salt.
  • Fluorinated C4 building block for synthesis.
  • CAS number 1780799-10-1.
  • Molecular weight 127.59 g/mol.
  • Purity 95.0% (as reported on certificate of analysis).
  • Solid; store sealed at room temperature, away from moisture and light.
  • Commonly supplied in small laboratory pack sizes such as 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.