Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorocarbazole 98.0+%, TCI America™
SDP

Catalog No. F09651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09651G Supplier TCI America Supplier No. F09651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 391-45-7
Molecular Formula C12H8FN
Molecular Weight (g/mol) 185.201
MDL Number MFCD18450178
InChI Key IEUMYICVZHBVAV-UHFFFAOYSA-N
PubChem CID 15886841
IUPAC Name 3-fluoro-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)F

Specifications

Melting Point 212°C
Quantity 1 g
Formula Weight 185.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.