Learn More
Medchemexpress LLC 3-Fluorocatechol | 363-52-0 | MFCD00042582 | 128.10 | 5 G
Supplier: Medchemexpress LLC HYW0074825G
3-Fluorocatechol is a fluorinated catechol primarily used as a biochemical reagent in laboratory settings for life science-related research and the manufacture of various substances. It serves as an intermediate in the preparation of synthetic humic acid. This product is intended for research use only and should be handled by experienced personnel in appropriately equipped facilities.
- Research-grade biochemical reagent
- Suitable for laboratory applications
- Precursor in synthetic humic acid production
- Identified as an inhibitor
- Part of screening libraries and protein research