Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. F08005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08005G 5 g
F080025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08005G Supplier TCI America Supplier No. F08005G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 701-83-7
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00142578
InChI Key IAWZWMGUTKRLQB-UHFFFAOYSA-N
Synonym 1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
PubChem CID 136542
IUPAC Name (3-fluorophenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)F

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorophenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.