Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenylacetone, 97%, Thermo Scientific™

Catalog No. AAB2206103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2206103 1 g
AAB2206106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2206103 Supplier Thermo Scientific Chemicals Supplier No. B2206103

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluorophenylacetone is used in the preparation of of optically active cyanohydrins. 3-Fluorophenylaceton is reactant in chiral amine synthesis.

Solubility
Soluble in chloroform, ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1737-19-5
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00061144
InChI Key UWCPYXSRCQVABG-UHFFFAOYSA-N
Synonym 3-fluorophenylacetone, 1-3-fluorophenyl propan-2-one, 1-3-fluorophenyl acetone, acmc-1bqff, m-fluorophenyl-2-propanone, 1-3-fluorophenyl acetone #, 1-3-fluorophenyl-2-propanone
PubChem CID 137185
IUPAC Name 1-(3-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)F

Specifications

Refractive Index 1.496
Quantity 1 g
Solubility Information Soluble in chloroform,ethyl Acetate.
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 3-Fluorophenylacetone
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.