Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific™

Catalog No. AAL2027803
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2027803 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2027803 Supplier Thermo Scientific Chemicals Supplier No. L2027803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 121247-01-6
Molecular Formula C8H7FO3
Quantity 1g
MDL Number MFCD05664101
Beilstein 3537863
Synonym 3-fluorophenylglyoxal hydrate, 2-3-fluorophenyl-2-oxoacetaldehyde hydrate, acmc-20an3a, 3-fluorophenyl glyoxal, 3-fluoro-a-oxobenzeneacetaldehyde, benzeneacetaldehyde,3-fluoro-a-oxo, 2-3-fluorophenyl-2-oxoethanal, hydrate
InChI Key VGFHAWFFIQFSRR-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)F)C(=O)C=O.O
IUPAC Name 2-(3-fluorophenyl)-2-oxoacetaldehyde;hydrate
Molecular Weight (g/mol) 170.139
PubChem CID 2783267
Formula Weight 152.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 3-Fluorophenylglyoxal hydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.