Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorophenylhydrazine hydrochloride, 98%

Catalog No. AAL0741806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0741806 5 g
AAL0741814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0741806 Supplier Thermo Scientific Chemicals Supplier No. L0741806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2924-16-5 | C6H8ClFN2 | 162.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2924-16-5
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.592
MDL Number MFCD00012934
InChI Key SKVGLOFWEJFQKU-UHFFFAOYSA-N
Synonym 3-fluorophenylhydrazine hydrochloride, 3-fluorophenyl hydrazine hydrochloride, 3-fluorophenylhydrazine hcl, 3-fluorophenyl hydrazine hcl, m-fluorophenylhydrazine hydrochloride, hydrazine, 3-fluorophenyl-, monohydrochloride, 1-3-fluorophenyl hydrazine hydrochloride, 3-fluoro-phenyl-hydrazine, pubchem3341, acmc-20a0dq
PubChem CID 2723914
IUPAC Name (3-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)F)NN.Cl

Specifications

Melting Point ∽268°C (decomposition)
Quantity 5 g
Beilstein 3627729
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 3-Fluorophenylhydrazine hydrochloride
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-886-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.