Learn More
AdipoGen 3-Fluorophthalic anhydride (25 g)
SDP
Supplier:  AdipoGen CDXF0148G025
Encompass_Preferred
Chemical. CAS: 652-39-1. Formula: C8H3FO3. MW: 166.11. Building block for synthesis. 3-Fluorophthalic anhydride is a useful fluorinated building block for the preparation of various compounds of interest in pharmaceutcials and agrochemicals.
Catalog No. 50-205-4445