Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoropyridine-4-carboxylic acid, 97%

Catalog No. AAL2004203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2004203 1 g
AAL2004206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2004203 Supplier Thermo Scientific Chemicals Supplier No. L2004203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 393-53-3 | C6H4FNO2 | 141.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-53-3
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD03789251
InChI Key POLXLLIQWDBJMD-UHFFFAOYSA-N
Synonym 3-fluoroisonicotinic acid, 3-fluoro-4-pyridinecarboxylic acid, 3-fluoro isonicotinic acid, 3-fluoro-isonicotinic acid, 3-fluoro-4-carboxypyridine, 4-pyridinecarboxylic acid, 3-fluoro, 3-fluoropyridine-4-carboxylicacid, pubchem3549, 3-fluoroisonicotinic acid?
PubChem CID 345361
IUPAC Name 3-fluoropyridine-4-carboxylic acid
SMILES C1=CN=CC(=C1C(=O)O)F

Specifications

Melting Point 251°C to 254°C (decomposition)
Quantity 1 g
Beilstein 383678
Formula Weight 141.1
Percent Purity 97%
Chemical Name or Material 3-Fluoropyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.