Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorosalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. F08451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08451G 1 g
F08455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08451G Supplier TCI America Supplier No. F08451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 341-27-5
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.112
MDL Number MFCD00153167
InChI Key GFHCXVJXSLGRJR-UHFFFAOYSA-N
Synonym 3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo
PubChem CID 268724
IUPAC Name 3-fluoro-2-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)F)O)C(=O)O

Specifications

Melting Point 148°C
Quantity 1 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorosalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.