Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Fluorosulfonyl)-2-thiophenecarboxylic acid methyl ester

Catalog No. ALD001681G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD001681G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD001681G Supplier Sigma Aldrich Supplier No. ALD001681G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 9-oxabicyclo[6.1.0]non-4-ene is an epoxide. it undergoes halofluorination reactions with n-halosuccinimides and triethylamine tris-hydrofluoride or olahαs reagent.9-oxabicyclo[6.1.0]non-4-ene may be used in the following studies:
  • Novel bicyclo[4.2.1] and bicyclo[3.3.1] ethers
  • Trans, trans-2,6-dibromo-9-oxabicyclo[3.3.1]nonane and trans, trans-2,5-dibromo-9-oxabicyclo[4.2.1]nonane
  • (z)-2-(cyclooct-4-enyloxy)acetic acid

Specifications

CAS 1373233-35-2
Melting Point 75°C to 80°C
Molecular Formula C6H5FO4S2
Linear Formula C6H5FO4S2
Quantity 5 g
Molecular Weight (g/mol) 224.23
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.