Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Fluorosulfonyl)benzoic acid

Catalog No. ALD000381G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD000381G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD000381G Supplier Sigma Aldrich Supplier No. ALD000381G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-methyl-p-toluidine is an aromatic secondary amine
  • It forms social isomers when encapsulated with a molecule of chloroform or benzene in a cylindrical capsule
  • 2-amino-4,6-dichloropyrimidine-5-carbaldehyde undergoes mono-amination with n-methyl-p-toluidine to form a precursor for the preparation of pyrazolo[3,4-d]pyrimidines
  • It annelates with dimethyl acetylenedicarboxylate via formation of toluidine radical cation intermediate

Specifications

Melting Point 147°C to 153°C
CAS 454-95-5
Molecular Formula C7H5FO4S
Linear Formula C7H5FO4S
MDL Number MFCD00007415
Synonym 3-(Fluorosulfonyl)benzoic acid
Molecular Weight (g/mol) 204.18
Quantity 25 mL
Percent Purity 95% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.