Learn More
3-Fluorothiophenol, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.203 |
| Boiling Point | 156°C |
| Flash Point | 58°C (136°F) |
| Odor | Stench |
| Refractive Index | 1.554 |
| Quantity | 5 g |
| UN Number | UN3071 |
| Beilstein | 2039302 |
| Sensitivity | Air Sensitive |
| Formula Weight | 128.17 |
| Show More |
Safety and Handling
| Hazard Category | H226-H301+H311+H331-H315-H319-H335 |
| Hazard Statement | GHS H Statement H300-H310-H331-H226-H315-H319-H335 Fatal if swallowed. Fatal in contact with skin. Toxic if inhaled. Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 219-876-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.