Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Formyl-6-isopropylchromone 98.0+%, TCI America™
SDP

Catalog No. F08561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08561G Supplier TCI America Supplier No. F08561G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 49619-58-1
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.24
MDL Number MFCD00192183
InChI Key FRRYMYQANNFABF-UHFFFAOYSA-N
Synonym 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde, 6-Isopropylchromone-3-carboxaldehyde
PubChem CID 688711
IUPAC Name 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbaldehyde
SMILES CC(C)C1=CC=C2OC=C(C=O)C(=O)C2=C1

Specifications

Melting Point 99°C
Color Yellow
Quantity 1 g
Formula Weight 216.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Formyl-6-isopropylchromone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.