Learn More
Cayman Chemical 3-formyl RIfamycIn 10g
SDP
Catalog No. 502983044
Encompass_Preferred
An intermediate of rifampicin that has been used to develop several rifamycin derivatives with antibiotic activity that targets RNA synthesis
Catalog No. 50-298-3044 Supplier Cayman Chemical Supplier No. 2026010 G
An intermediate of rifampicin that has been used to develop several rifamycin derivatives with antibiotic activity that targets RNA synthesis