Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Formyl rifamycin | 13292-22-3 | 98.5% | 1 ML
SDP

Catalog No. 5000338848
Encompass_Preferred

3-Formyl rifamycin is an intermediate of Rifampicin, developed for research applications. It presents as a pink to red solid, with a molecular weight of 725.78 and a chemical formula of C38H47NO13. This compound is soluble in DMSO at 62.5 mg/mL.

  • Intermediate of Rifampicin
  • Intended for research use only
  • Appears as a pink to red solid
  • Molecular weight: 725.78
  • Chemical formula: C38H47NO13
  • Soluble in DMSO

Catalog No. 50-003-38848 Supplier Medchemexpress LLC Supplier No. HY12904310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Formyl rifamycin is an intermediate of Rifampicin, developed for research applications. It presents as a pink to red solid, with a molecular weight of 725.78 and a chemical formula of C38H47NO13. This compound is soluble in DMSO at 62.5 mg/mL.

  • Intermediate of Rifampicin
  • Intended for research use only
  • Appears as a pink to red solid
  • Molecular weight: 725.78
  • Chemical formula: C38H47NO13
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.