Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Formylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
87199-16-4
C7H7BO3
149.94
MFCD00161356
HJBGZJMKTOMQRR-UHFFFAOYSA-N
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
2734356
(3-formylphenyl)boronic acid
OB(O)C1=CC=CC(C=O)=C1
87199-16-4
C7H7BO3
149.94
MFCD00161356
HJBGZJMKTOMQRR-UHFFFAOYSA-N
3-formylphenyl boronic acid, 3-formylbenzeneboronic acid, 3-boronobenzaldehyde, 3-formyl phenylboronic acid, 3-formylboronic acid, m-formylphenylboronic acid, 3-dihydroxyboryl benzaldehyde, boronic acid, 3-formylphenyl, 3-formylphenylboronicacid
2734356
(3-formylphenyl)boronic acid
OB(O)C1=CC=CC(C=O)=C1
Melting Point 100°C
Packaging Glass bottle
Quantity 1g
Color Beige to White or Orange
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 149.94
Physical Form Powder, Crystals, and/or Chunks
Percent Purity 97%
Chemical Name or Material 3-Formylphenylboronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning