Learn More
Lkt Laboratories 3-Formylrifamycin, ≥98%, 1GR
SDP
Supplier:  Lkt Laboratories R32211G
Encompass_Preferred
Rifamycin derivative, induces pore formation in mitochondrial membranes; DNA-dependent RNA polymerase inhibitor. CAS: 13292-22-3. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4963