Learn More
Cayman Chemical 3-FormylsalIcylIc AcId 10mg
SDP
Catalog No. 502994781
Encompass_Preferred
An inhibitor of estrone sulfatase (IC50 = 150 nM; Ki = 120 nM); has been used in the synthesis of Schiff bases
Catalog No. 50-299-4781 Supplier Cayman Chemical Supplier No. 2336810 MG
An inhibitor of estrone sulfatase (IC50 = 150 nM; Ki = 120 nM); has been used in the synthesis of Schiff bases