Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Formylsalicylic Acid 97%, Thermo Scientific™

Catalog No. AC276480010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC276480010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC276480010 Supplier Thermo Scientific Chemicals Supplier No. 276480010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-04-8
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00143165
InChI Key YOEUNKPREOJHBW-UHFFFAOYSA-N
Synonym 3-formylsalicylic acid, 2-hydroxyisophthalaldehydic acid, isophthalaldehydic acid, 2-hydroxy, 2-hydroxy-3-formylbenzoic acid, 3-carboxy-2-hydroxybenzaldehyde, 3-formylsalicylic acid,97, carboxy-2-hydroxybenzaldehyde, yoeunkpreojhbw-uhfffaoysa, benzoic acid,3-formyl-2-hydroxy, benzoic acid, 3-formyl-2-hydroxy
PubChem CID 69117
ChEBI CHEBI:34334
IUPAC Name 3-formyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(C=O)=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 175°C to 180°C
Color Beige to Brown
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 166.13
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 3-Formylsalicylic acid, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.