Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Furaldehyde | 498-60-2 | 99.9% | C5H4O2 | 1 G
SDP

Catalog No. 5000434157
Encompass_Preferred

3-Furaldehyde is a furan aldehyde used in the synthesis of the neoclerodane diterpene Salvinorin A. It is also recognized as an atmospheric pollutant, predominantly resulting from biomass burning, and is known to undergo photoisomerization and photolysis reactions under ultraviolet light.

  • Used in the synthesis of Salvinorin A
  • Recognized as an atmospheric pollutant
  • Undergoes photoisomerization and photolysis under UV light
  • Appears as a colorless to light yellow liquid
  • Purity is 99.9%
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-34157 Supplier Medchemexpress LLC Supplier No. HY762241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Furaldehyde is a furan aldehyde used in the synthesis of the neoclerodane diterpene Salvinorin A. It is also recognized as an atmospheric pollutant, predominantly resulting from biomass burning, and is known to undergo photoisomerization and photolysis reactions under ultraviolet light.

  • Used in the synthesis of Salvinorin A
  • Recognized as an atmospheric pollutant
  • Undergoes photoisomerization and photolysis under UV light
  • Appears as a colorless to light yellow liquid
  • Purity is 99.9%
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.