Learn More
Medchemexpress LLC 3-Furaldehyde | 498-60-2 | 99.9% | C5H4O2 | 10 G
SDP
Catalog No. 5000434110
Encompass_Preferred
3-Furaldehyde is a furan and an aldehyde compound utilized in the synthesis of Salvinorin A and is an atmospheric pollutant primarily originating from biomass burning. It undergoes photoisomerization and photolysis reactions under ultraviolet light, leading to various chemical transformations. This product is intended for research use only.
  • Categorized as a furan and an aldehyde
  • Can be used in the synthesis of Salvinorin A
  • Undergoes photoisomerization reaction under ultraviolet light
  • Undergoes photolysis reaction under higher energy ultraviolet light
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
Catalog No. 50-004-34110 Supplier Medchemexpress LLC Supplier No. HY7622410G
3-Furaldehyde is a furan and an aldehyde compound utilized in the synthesis of Salvinorin A and is an atmospheric pollutant primarily originating from biomass burning. It undergoes photoisomerization and photolysis reactions under ultraviolet light, leading to various chemical transformations. This product is intended for research use only.
  • Categorized as a furan and an aldehyde
  • Can be used in the synthesis of Salvinorin A
  • Undergoes photoisomerization reaction under ultraviolet light
  • Undergoes photolysis reaction under higher energy ultraviolet light
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation