Learn More
Medchemexpress LLC 3-Furaldehyde | 498-60-2 | 99.94% | C5H4O2 | 5 G
SDP
Catalog No. 5000434159
Encompass_Preferred
3-Furaldehyde is a furan member and an aldehyde used in synthesizing the neoclerodane diterpene Salvinorin A. It is also an atmospheric pollutant from biomass burning, undergoing photoisomerization and photolysis under ultraviolet light. It is available as a colorless to light yellow liquid.
  • Can be used in the synthesis of Salvinorin A
  • Atmospheric pollutant derived from biomass burning
  • Undergoes photoisomerization and photolysis reactions
  • Research use only
Catalog No. 50-004-34159 Supplier Medchemexpress LLC Supplier No. HY762245G
3-Furaldehyde is a furan member and an aldehyde used in synthesizing the neoclerodane diterpene Salvinorin A. It is also an atmospheric pollutant from biomass burning, undergoing photoisomerization and photolysis under ultraviolet light. It is available as a colorless to light yellow liquid.
  • Can be used in the synthesis of Salvinorin A
  • Atmospheric pollutant derived from biomass burning
  • Undergoes photoisomerization and photolysis reactions
  • Research use only