Learn More
Medchemexpress LLC 3-Furaldehyde | 498-60-2 | 99.98% | C5H4O2 | 25 G
SDP
Catalog No. 5000357617
Encompass_Preferred
3-Furaldehyde is an aldehyde and a member of furans. It is used in the synthesis of the neoclerodane diterpene Salvinorin A and is intended for research use only. This compound is also recognized as an atmospheric pollutant, primarily originating from biomass burning, and exhibits photoisomerization and photolysis reactions when exposed to ultraviolet light.
  • Undergoes photoisomerization, converting *trans* to *cis* isomer under UV light.
  • Exhibits photolysis reaction, leading to decarbonylation to form furan, cyclopropene, and acrylene under higher UV energy.
  • Essential for synthesizing the neoclerodane diterpene Salvinorin A.
  • Identified as an atmospheric pollutant derived from biomass burning.
Catalog No. 50-003-57617 Supplier Medchemexpress LLC Supplier No. 25GHY76224
3-Furaldehyde is an aldehyde and a member of furans. It is used in the synthesis of the neoclerodane diterpene Salvinorin A and is intended for research use only. This compound is also recognized as an atmospheric pollutant, primarily originating from biomass burning, and exhibits photoisomerization and photolysis reactions when exposed to ultraviolet light.
  • Undergoes photoisomerization, converting *trans* to *cis* isomer under UV light.
  • Exhibits photolysis reaction, leading to decarbonylation to form furan, cyclopropene, and acrylene under higher UV energy.
  • Essential for synthesizing the neoclerodane diterpene Salvinorin A.
  • Identified as an atmospheric pollutant derived from biomass burning.