Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Furaldehyde, 98%, stabilized

Catalog No. ac393260010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393260010 1 g Glass bottle
AC393260050 5 g Glass bottle
AC393260250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC393260010 Supplier Thermo Scientific Chemicals Supplier No. 393260010
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 498-60-2 | C5H4O2 | 96.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-60-2
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.08
InChI Key AZVSIHIBYRHSLB-UHFFFAOYSA-N
Synonym 3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
PubChem CID 10351
ChEBI CHEBI:87609
IUPAC Name furan-3-carbaldehyde
SMILES C1=COC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1500g/mL
Boiling Point 145.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.15
Formula Weight 96.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Furaldehyde
Show More Show Less
Hazard Category GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.- No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathi
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.